missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichlorodiethylsilane 98.0+%, TCI America™
Supplier: TCI America D19765G
Specifications
| Dichlorodiethylsilane | |
| -97°C | |
| 129°C | |
| MFCD00000493 | |
| 1767 | |
| BYLOHCRAPOSXLY-UHFFFAOYSA-N | |
| dichlorodiethylsilane | |
| 15589 | |
| ≥98.0% (GC) |
| 1719-53-5 | |
| Yellow | |
| C4H10Cl2Si | |
| 5 g | |
| diethyldichlorosilane, silane, dichlorodiethyl, dichloro diethyl silane, diethyldichlorosilicon, diethyl dichlorosilane, diethyidichlorosilane, snpladqptpdumup@, dichlorodiethyl-silan, diethylsiliciumdichlorid, silane,dichlorodiethyl | |
| CC[Si](Cl)(Cl)CC | |
| 157.11 | |
| 157.11 | |
| Liquid |
Chemical Identifiers
| 1719-53-5 | |
| 157.11 | |
| BYLOHCRAPOSXLY-UHFFFAOYSA-N | |
| 15589 | |
| CC[Si](Cl)(Cl)CC |
| C4H10Cl2Si | |
| MFCD00000493 | |
| diethyldichlorosilane, silane, dichlorodiethyl, dichloro diethyl silane, diethyldichlorosilicon, diethyl dichlorosilane, diethyidichlorosilane, snpladqptpdumup@, dichlorodiethyl-silan, diethylsiliciumdichlorid, silane,dichlorodiethyl | |
| dichlorodiethylsilane |
Safety and Handling
EINECSNumber : (2)-2041
RTECSNumber : VV3060000
TSCA : Yes