missing translation for 'onlineSavingsMsg'
Learn More

Dichlorodiethylsilane 98.0+%, TCI America™

Catalog No. p-7127042
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
D19765G 5 g
D197625G 25 g
2 options

Catalog No. D19765G

Supplier: TCI America D19765G

Only null left

Chemical Identifiers

CAS 1719-53-5
Molecular Formula C4H10Cl2Si
Molecular Weight (g/mol) 157.11
MDL Number MFCD00000493
InChI Key BYLOHCRAPOSXLY-UHFFFAOYSA-N
Synonym diethyldichlorosilane, silane, dichlorodiethyl, dichloro diethyl silane, diethyldichlorosilicon, diethyl dichlorosilane, diethyidichlorosilane, snpladqptpdumup@, dichlorodiethyl-silan, diethylsiliciumdichlorid, silane,dichlorodiethyl
PubChem CID 15589
IUPAC Name dichlorodiethylsilane
SMILES CC[Si](Cl)(Cl)CC

Specifications

Melting Point -97°C
Color Yellow
Boiling Point 129°C
Quantity 5 g
UN Number 1767
Formula Weight 157.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dichlorodiethylsilane
EINECSNumber (2)-2041
RTECSNumber VV3060000
TSCA Yes