missing translation for 'onlineSavingsMsg'
Learn More

Dichlorobis(tricyclohexylphosphine)palladium(II), 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 395890010

 View more versions of this product

Catalog No. AC395890010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Dichlorobis(tricyclohexylphosphine)palladium(II)
97.5% min.
98%
C36H66Cl2P2Pd
1g
VUYVXCJTTQJVKJ-UHFFFAOYSA-L
dichloropalladium;tricyclohexylphosphane
11050900
98%
29934-17-6
Authentic
Glass bottle
MFCD00191830
dichlorobis tricyclohexylphosphine palladium ii, bis tricyclohexylphosphine palladium ii dichloride, dichlorobis tricyclohexylphosphine palladium, bis tricyclohexylphosphine-palladium ii dichloride, bis tricyclohexylphosphine palladium ii-dichloride, bis tricyclo-hexylphosphine palladium ii dichloride, bis tricyclohexylphosphine palladium ii chloride
C1CCC(CC1)P(C2CCCCC2)C3CCCCC3.C1CCC(CC1)P(C2CCCCC2)C3CCCCC3.Cl[Pd]Cl
738.18
738.18

Chemical Identifiers

29934-17-6
738.18
VUYVXCJTTQJVKJ-UHFFFAOYSA-L
11050900
C1CCC(CC1)P(C2CCCCC2)C3CCCCC3.C1CCC(CC1)P(C2CCCCC2)C3CCCCC3.Cl[Pd]Cl
C36H66Cl2P2Pd
MFCD00191830
dichlorobis tricyclohexylphosphine palladium ii, bis tricyclohexylphosphine palladium ii dichloride, dichlorobis tricyclohexylphosphine palladium, bis tricyclohexylphosphine-palladium ii dichloride, bis tricyclohexylphosphine palladium ii-dichloride, bis tricyclo-hexylphosphine palladium ii dichloride, bis tricyclohexylphosphine palladium ii chloride
dichloropalladium;tricyclohexylphosphane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only