missing translation for 'onlineSavingsMsg'
Learn More

Dichlorobis(ethylenediamine)platinum(II), Pt 50%

CAS: 21430-85-3 | C4H16Cl2N4Pt | 386.18 g/mol

$150.93 - $550.78

Chemical Identifiers

CAS 21430-85-3
Molecular Formula C4H16Cl2N4Pt
Molecular Weight (g/mol) 386.18
MDL Number MFCD00009881
InChI Key MXLIXEZAMQLDMM-UHFFFAOYSA-L
Synonym Bis(ethylenediamine)platinum(II) chloride
IUPAC Name platinum(2+) bis(ethane-1,2-diamine) dichloride
SMILES [Cl-].[Cl-].[Pt++].NCCN.NCCN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3928001
View Documents
Thermo Scientific Chemicals
03928001
0.25 g
Each for $150.93
Only null left
 
AA3928003
View Documents
Thermo Scientific Chemicals
03928003
1 g
Each for $550.78
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

21430-85-3
386.18
MXLIXEZAMQLDMM-UHFFFAOYSA-L
platinum(2+) bis(ethane-1,2-diamine) dichloride
C4H16Cl2N4Pt
MFCD00009881
Bis(ethylenediamine)platinum(II) chloride
[Cl-].[Cl-].[Pt++].NCCN.NCCN

Specifications

21430-85-3
White
PtCl2(H2NCH2CH2NH2)2
0.25 g
MXLIXEZAMQLDMM-UHFFFAOYSA-L
platinum(2+) bis(ethane-1,2-diamine) dichloride
386.18
Crystalline
300°C (decomposition)
C4H16Cl2N4Pt
MFCD00009881
Bis(ethylenediamine)platinum(II) chloride
[Cl-].[Cl-].[Pt++].NCCN.NCCN
386.18
Pt 50%
Dichlorobis(ethylenediamine)platinum(II)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c

H315-H317-H319-H334-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only