accessibility menu, dialog, popup

UserName

NNNNPtClCl

CAS RN 21430-85-3

IUPAC Name: platinum(2+) bis(ethane-1,2-diamine) dichloride
Synonyms: platinum(2+) bis(ethylenediamine) dichloride
Molecular Weight (g/mol): 386.18
Molecular Formula: C4H16Cl2N4Pt
InChi Key: MXLIXEZAMQLDMM-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Pt++].NCCN.NCCN
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results