missing translation for 'onlineSavingsMsg'
Learn More

Dichlorobis(ethylenediamine)platinum(II), Pt 50%

Catalog No. AA3928001
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3928001 0.25 g
AA3928003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3928001 Supplier Thermo Scientific Chemicals Supplier No. 03928001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 21430-85-3 | C4H16Cl2N4Pt | 386.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21430-85-3
Molecular Formula C4H16Cl2N4Pt
Molecular Weight (g/mol) 386.18
MDL Number MFCD00009881
InChI Key MXLIXEZAMQLDMM-UHFFFAOYSA-L
Synonym Bis(ethylenediamine)platinum(II) chloride
IUPAC Name platinum(2+) bis(ethane-1,2-diamine) dichloride
SMILES [Cl-].[Cl-].[Pt++].NCCN.NCCN

Specifications

Melting Point 300°C (decomposition)
Color White
Linear Formula PtCl2(H2NCH2CH2NH2)2
Quantity 0.25 g
Formula Weight 386.18
Concentration or Composition (by Analyte or Components) Pt 50%
Physical Form Crystalline
Chemical Name or Material Dichlorobis(ethylenediamine)platinum(II)
Hazard Category H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only