missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloroacetonitrile 98.0+%, TCI America™
$156.59 - $1865.92
Chemical Identifiers
| CAS | 3018-12-0 |
|---|---|
| Molecular Formula | C2HCl2N |
| Molecular Weight (g/mol) | 109.937 |
| MDL Number | MFCD00040886 |
| InChI Key | STZZWJCGRKXEFF-UHFFFAOYSA-N |
| Synonym | dichloroacetonitrile, acetonitrile, dichloro, dichloromethyl cyanide, ccris 942, unii-o0l64v63m9, dichloro acetonitrile, pubchem17242, acmc-209hdn, dsstox_cid_1562 |
| PubChem CID | 18177 |
| ChEBI | CHEBI:82444 |
| IUPAC Name | 2,2-dichloroacetonitrile |
| SMILES | C(#N)C(Cl)Cl |
Chemical Identifiers
| 3018-12-0 | |
| 109.937 | |
| STZZWJCGRKXEFF-UHFFFAOYSA-N | |
| 18177 | |
| 2,2-dichloroacetonitrile |
| C2HCl2N | |
| MFCD00040886 | |
| dichloroacetonitrile, acetonitrile, dichloro, dichloromethyl cyanide, ccris 942, unii-o0l64v63m9, dichloro acetonitrile, pubchem17242, acmc-209hdn, dsstox_cid_1562 | |
| CHEBI:82444 | |
| C(#N)C(Cl)Cl |
Specifications
| 3018-12-0 | |
| 112°C | |
| MFCD00040886 | |
| 2920 | |
| STZZWJCGRKXEFF-UHFFFAOYSA-N | |
| 2,2-dichloroacetonitrile | |
| 18177 | |
| 109.94 | |
| Liquid |
| Yellow | |
| C2HCl2N | |
| 25 g | |
| dichloroacetonitrile, acetonitrile, dichloro, dichloromethyl cyanide, ccris 942, unii-o0l64v63m9, dichloro acetonitrile, pubchem17242, acmc-209hdn, dsstox_cid_1562 | |
| C(#N)C(Cl)Cl | |
| 109.937 | |
| CHEBI:82444 | |
| ≥98.0% (GC) | |
| Dichloroacetonitrile |
Safety and Handling
RTECSNumber : AL8465000
TSCA : No