accessibility menu, dialog, popup

UserName

ClClN

CAS RN 3018-12-0

IUPAC Name: 2,2-dichloroacetonitrile
Synonyms: dichloroacetonitrile
Molecular Weight (g/mol): 109.94
Molecular Formula: C2HCl2N
InChi Key: STZZWJCGRKXEFF-UHFFFAOYSA-N
SMILES: ClC(Cl)C#N
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)