missing translation for 'onlineSavingsMsg'
Learn More

Dichloroacetic Acid 98.0+%, TCI America™

$123.03 - $123.03

Chemical Identifiers

CAS 79-43-6
Molecular Formula C2H2Cl2O2
Molecular Weight (g/mol) 128.936
MDL Number MFCD00004223
InChI Key JXTHNDFMNIQAHM-UHFFFAOYSA-N
Synonym dichloroacetic acid, dichloracetic acid, dichloroethanoic acid, acetic acid, dichloro, urner's liquid, dichlorethanoic acid, bichloracetic acid, dca acid, bichloroacetic acid, dichloro-acetic acid
PubChem CID 6597
ChEBI CHEBI:36386
IUPAC Name 2,2-dichloroacetic acid
SMILES C(C(=O)O)(Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D030825G
View Documents
TCI America
D030825G
25 g
Each for $123.03
Only null left
 
Description

Description

Specifications

Chemical Identifiers

79-43-6
128.936
JXTHNDFMNIQAHM-UHFFFAOYSA-N
6597
2,2-dichloroacetic acid
C2H2Cl2O2
MFCD00004223
dichloroacetic acid, dichloracetic acid, dichloroethanoic acid, acetic acid, dichloro, urner's liquid, dichlorethanoic acid, bichloracetic acid, dca acid, bichloroacetic acid, dichloro-acetic acid
CHEBI:36386
C(C(=O)O)(Cl)Cl

Specifications

79-43-6
193°C
MFCD00004223
1764
JXTHNDFMNIQAHM-UHFFFAOYSA-N
2,2-dichloroacetic acid
6597
128.94
Liquid
1.2 (129g/L H2O soln.)
C2H2Cl2O2
25 g
dichloroacetic acid, dichloracetic acid, dichloroethanoic acid, acetic acid, dichloro, urner's liquid, dichlorethanoic acid, bichloracetic acid, dca acid, bichloroacetic acid, dichloro-acetic acid
C(C(=O)O)(Cl)Cl
128.936
CHEBI:36386
≥98.0% (GC,T)
Dichloroacetic Acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1161

RTECSNumber : AG6125000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'