missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloro(p-cymene)ruthenium(II) Dimer 95.0+%, TCI America™
$115.82 - $558.20
Chemical Identifiers
| CAS | 52462-29-0 |
|---|---|
| Molecular Formula | C20H28Cl4Ru2 |
| Molecular Weight (g/mol) | 612.384 |
| MDL Number | MFCD00064793 |
| InChI Key | LAXRNWSASWOFOT-UHFFFAOYSA-J |
| Synonym | dichloro p-cymene ruthenium ii dimer, di-mu-chloro-bis chloro p-cymene ruthenium ii, rucl2 p-cymene 2, ru p-cymene cl2 2, dichloro p-cymene ruthenium dimer, p-cymene ruthenium ii chloride dimer, dichloro-p-cymene ruthenium ii dimer |
| PubChem CID | 10908223 |
| IUPAC Name | dichlororuthenium;1-methyl-4-propan-2-ylbenzene |
| SMILES | CC1=CC=C(C=C1)C(C)C.CC1=CC=C(C=C1)C(C)C.Cl[Ru]Cl.Cl[Ru]Cl |
Chemical Identifiers
| 52462-29-0 | |
| 612.384 | |
| LAXRNWSASWOFOT-UHFFFAOYSA-J | |
| 10908223 | |
| CC1=CC=C(C=C1)C(C)C.CC1=CC=C(C=C1)C(C)C.Cl[Ru]Cl.Cl[Ru]Cl |
| C20H28Cl4Ru2 | |
| MFCD00064793 | |
| dichloro p-cymene ruthenium ii dimer, di-mu-chloro-bis chloro p-cymene ruthenium ii, rucl2 p-cymene 2, ru p-cymene cl2 2, dichloro p-cymene ruthenium dimer, p-cymene ruthenium ii chloride dimer, dichloro-p-cymene ruthenium ii dimer | |
| dichlororuthenium;1-methyl-4-propan-2-ylbenzene |
Specifications
| 52462-29-0 | |
| MFCD00064793 | |
| LAXRNWSASWOFOT-UHFFFAOYSA-J | |
| dichlororuthenium;1-methyl-4-propan-2-ylbenzene | |
| 10908223 | |
| ≥95.0% (T) | |
| Dichloro(p-cymene)ruthenium(II) Dimer |
| C20H28Cl4Ru2 | |
| dichloro p-cymene ruthenium ii dimer, di-mu-chloro-bis chloro p-cymene ruthenium ii, rucl2 p-cymene 2, ru p-cymene cl2 2, dichloro p-cymene ruthenium dimer, p-cymene ruthenium ii chloride dimer, dichloro-p-cymene ruthenium ii dimer | |
| CC1=CC=C(C=C1)C(C)C.CC1=CC=C(C=C1)C(C)C.Cl[Ru]Cl.Cl[Ru]Cl | |
| 612.384 | |
| 612.38 | |
| Crystalline Powder |
Safety and Handling
TSCA : No