missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(p-cymene)ruthenium(II) Dimer 95.0+%, TCI America™

Catalog No. D27515G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27515G 5 g
D27511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27515G Supplier TCI America Supplier No. D27515G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 52462-29-0
Molecular Formula C20H28Cl4Ru2
Molecular Weight (g/mol) 612.384
MDL Number MFCD00064793
InChI Key LAXRNWSASWOFOT-UHFFFAOYSA-J
Synonym dichloro p-cymene ruthenium ii dimer, di-mu-chloro-bis chloro p-cymene ruthenium ii, rucl2 p-cymene 2, ru p-cymene cl2 2, dichloro p-cymene ruthenium dimer, p-cymene ruthenium ii chloride dimer, dichloro-p-cymene ruthenium ii dimer
PubChem CID 10908223
IUPAC Name dichlororuthenium;1-methyl-4-propan-2-ylbenzene
SMILES CC1=CC=C(C=C1)C(C)C.CC1=CC=C(C=C1)C(C)C.Cl[Ru]Cl.Cl[Ru]Cl

Specifications

Quantity 5 g
Formula Weight 612.38
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dichloro(p-cymene)ruthenium(II) Dimer