missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(methyl)octadecylsilane 94.0+%, TCI America™

Catalog No. O026425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O026425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O026425ML Supplier TCI America Supplier No. O026425ML
Only null left

Chemical Identifiers

CAS 5157-75-5
Molecular Formula C19H40Cl2Si
Molecular Weight (g/mol) 367.51
MDL Number MFCD00018083
InChI Key GYWBHBXGYTYXRG-UHFFFAOYSA-N
Synonym methyloctadecyldichlorosilane, stearylmethyldichlorosilane, dichloro methyl octadecylsilane, silane, dichloromethyloctadecyl, dichloromethyloctadecylsilane, n-octadecylmethyldichlorosilane, dichloro methyl octadecyl silane, silane odm, acmc-209kum, dichlorooctadecylmethylsilane
PubChem CID 78835
IUPAC Name dichloro(methyl)octadecylsilane
SMILES CCCCCCCCCCCCCCCCCC[Si](C)(Cl)Cl

Specifications

Color White
Boiling Point 210°C
Quantity 25 mL
UN Number 2987
Formula Weight 367.51
Percent Purity ≥94.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Dichloro(methyl)octadecylsilane
EINECSNumber (2)-2041
TSCA Yes