CAS RN 5157-75-5
CAS RN 5157-75-5
IUPAC Name:
dichloro(methyl)octadecylsilane
Synonyms:
dichloro(methyl)octadecylsilane
Molecular Weight (g/mol):
367.51
Molecular Formula:
C19H40Cl2Si
InChi Key:
GYWBHBXGYTYXRG-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCC[Si](C)(Cl)Cl
1
–
1
of
1
results
1
Dichloro(methyl)octadecylsilane 94.0+%, TCI America™
CAS: 5157-75-5 Molecular Formula: C19H40Cl2Si Molecular Weight (g/mol): 367.51 MDL Number: MFCD00018083 InChI Key: GYWBHBXGYTYXRG-UHFFFAOYSA-N Synonym: methyloctadecyldichlorosilane,stearylmethyldichlorosilane,dichloro methyl octadecylsilane,silane, dichloromethyloctadecyl,dichloromethyloctadecylsilane,n-octadecylmethyldichlorosilane,dichloro methyl octadecyl silane,silane odm,acmc-209kum,dichlorooctadecylmethylsilane PubChem CID: 78835 IUPAC Name: dichloro(methyl)octadecylsilane SMILES: CCCCCCCCCCCCCCCCCC[Si](C)(Cl)Cl