missing translation for 'onlineSavingsMsg'
Learn More

Dichloro[bis(1,4-diphenylphosphino)butane]palladium(II), Pd 17.6%

Catalog No. p-7059600
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4497103 1 g
AA4497106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4497103 Supplier Thermo Scientific Chemicals Supplier No. 04497103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 29964-62-3 | C28H28Cl2P2Pd | 603.80 g/mol

Dichloro[bis(1,4-diphenylphosphino)butane]palladium(II) is used for the catalysis of styrene carbonylation, coupling of alkyl Grignard reagents with organic halides, selective monoalkylation of organic polyhalides, and modification of the dihalovinyl moiety of synthetic pyrethroids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29964-62-3
Molecular Formula C28H28Cl2P2Pd
Molecular Weight (g/mol) 603.80
MDL Number MFCD02093437
InChI Key JQXJBXVWVPVTOO-UHFFFAOYSA-L
Synonym 1,4-bis diphenylphosphino butane-palladium ii chloride, 1,4-butylenebis diphenylphosphine palladium dichloride, pdcl2 dppb, 1,4-bis diphenylphosphino butane palladium ii dichloride, palladium ii chloride-1,4-bis diphenylphosphine butane, dichloro bis 1,4-diphenylphosphino butane palladium ii, palladium ii chloride 1,4-bis diphenylphosphino butane complex, dichloro 1,4-bis diphenylphosphino butane palladium ii, pd dppb cl2, acmc-209sk1
PubChem CID 10974057
IUPAC Name dichloropalladium;4-diphenylphosphanylbutyl(diphenyl)phosphane
SMILES Cl[Pd]Cl.C(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 1 g
Solubility Information Soluble in ethanol,benzene,and ethereal solvents.
Formula Weight 603.8
Concentration or Composition (by Analyte or Components) Pd 17.6%
Physical Form Lumps
Chemical Name or Material Dichloro[bis (1,4-diphenylphosphino)butane]palladium (II)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only