accessibility menu, dialog, popup

UserName

CAS RN 29964-62-3

IUPAC Name: [4-(diphenylphosphanyl)butyl]diphenylphosphane; dichloropalladium
Synonyms: [4-(diphenylphosphanyl)butyl]diphenylphosphane; palladium chloride
Molecular Weight (g/mol): 603.8
Molecular Formula: C28H28Cl2P2Pd
InChi Key: JQXJBXVWVPVTOO-UHFFFAOYSA-L
SMILES: Cl[Pd]Cl.C(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)