missing translation for 'onlineSavingsMsg'
Learn More

Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II) 98.0+%, TCI America™

Catalog No. p-7128228
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4333200MG 200 mg
D43331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4333200MG Supplier TCI America Supplier No. D4333200MG
Only null left

Chemical Identifiers

CAS 205319-10-4
Molecular Formula C39H32Cl2OP2Pd
Molecular Weight (g/mol) 755.95
MDL Number MFCD14155707
InChI Key MNBAPNBMBOYNAW-UHFFFAOYSA-L
Synonym dichloro 9,9-dimethyl-4,5-bis diphenylphosphino xanthene palladium ii, palladium 2+ xantphos dichloride, 4,5-bis diphenylphosphino-9,9-dimethylxanthene dichloropalladium ii, 4,5-bis diphenylphosphino-9,9-dimethylxanthene palladium ii dichloride
PubChem CID 86280406
IUPAC Name [5-(diphenylphosphanyl)-9,9-dimethyl-9H-xanthen-4-yl]diphenylphosphane; dichloropalladiumbis(ylium)
SMILES Cl[Pd++]Cl.CC1(C)C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 287°C
Quantity 200 mg
Formula Weight 755.95
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II)