missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloramine B 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 74°C |
| Quantity | 5 g |
| UN Number | 1479 |
| Formula Weight | 226.07 |
| Percent Purity | ≥95.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Dichloramine B |
Safety and Handling
| EINECSNumber | (3)-3131 |
| TSCA | Yes |