missing translation for 'onlineSavingsMsg'
Learn More

Dicarbonylbis(cyclopentadienyl)titanium(II), 98%

Catalog No. AA3957102
Change view
Click to view available options
Quantity:
0.5 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3957102 0.5 g
AA3957104 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3957102 Supplier Thermo Scientific Chemicals Supplier No. 03957102

CAS: 12129-51-0 | C12H12O2Ti | 236.09 g/mol

It is used as an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12129-51-0
Molecular Formula C12H12O2Ti
Molecular Weight (g/mol) 236.09
MDL Number MFCD00143762
InChI Key AGUCYLATRKJGHM-UHFFFAOYSA-N
Synonym dicarbonylbis cyclopentadienyl titanium ii, acmc-1bra9, bis oxomethylidene titanium;cyclopenta-1,3-diene

Specifications

Melting Point 90°C (decomposition)
Boiling Point 60°C (0.001 mmHg)(sublimation)
Quantity 0.5 g
UN Number UN3466
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 234.09
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Dicarbonylbis(cyclopentadienyl)titanium(II)
Hazard Category H228-H301-H332
Hazard Statement GHS H Statement
H228-H300-H331
Flammable solid.
Fatal if swallowed.
Toxic if inhaled.
Precautionary Statement P210-P240-P241-P261-P264b-P270-P271-P280-P301+P310-P304+P340-P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only