accessibility menu, dialog, popup

UserName

CAS RN 12129-51-0

IUPAC Name: 1,1-bis(hydroxymethyl)-1-titana-1,1'-spirobi[pentacyclo[2.2.0.0¹,³.0¹,⁵.0²,⁶]hexane]
Synonyms: 1,1-bis(hydroxymethyl)-1-titana-1,1'-spirobi[pentacyclo[2.2.0.0¹,³.0¹,⁵.0²,⁶]hexane]
Molecular Weight (g/mol): 236.09
Molecular Formula: C12H12O2Ti
InChi Key: AGUCYLATRKJGHM-UHFFFAOYSA-N
SMILES: [H][O]#C[Ti]123456789(C#[O][H])C%10C1C2C3C4%10.C51C6C7C8C91
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)