Learn More
Dicarbonylacetylacetonato rhodium(I)
CAS: 14874-82-9 | C7H7O4Rh | 258.03 g/mol
$1129.62 - $5068.81
Chemical Identifiers
| CAS | 14874-82-9 |
|---|---|
| Molecular Formula | C7H7O4Rh |
| Molecular Weight (g/mol) | 258.03 |
| MDL Number | MFCD00009884 |
| InChI Key | BZCAWKOPWNIDOC-FGSKAQBVSA-M |
| IUPAC Name | λ¹-rhodium(1+) bis(methanidylidyneoxidanium) (2Z)-4-oxopent-2-en-2-olate |
| SMILES | [Rh+].[C-]#[O+].[C-]#[O+].C\C([O-])=C\C(C)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC363955000
|
Thermo Scientific Chemicals
363955000 |
500 mg | Glass bottle |
Each for $1,129.62
|
|
||||
|
AC363950025
|
Thermo Scientific Chemicals
363950025 |
2.5 g | Glass bottle |
Each for $5,068.81
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 14874-82-9 | |
| 258.03 | |
| BZCAWKOPWNIDOC-FGSKAQBVSA-M | |
| [Rh+].[C-]#[O+].[C-]#[O+].C\C([O-])=C\C(C)=O |
| C7H7O4Rh | |
| MFCD00009884 | |
| λ¹-rhodium(1+) bis(methanidylidyneoxidanium) (2Z)-4-oxopent-2-en-2-olate |
Specifications
| 14874-82-9 | |
| Green-Red | |
| Glass bottle | |
| MFCD00009884 | |
| Solubility in water: insoluble | |
| [Rh+].[C-]#[O+].[C-]#[O+].C\C([O-])=C\C(C)=O | |
| 258.03 | |
| Flakes or Needles |
| 154.0°C to 156.0°C | |
| Authentic | |
| C7H7O4Rh | |
| 500 mg | |
| BZCAWKOPWNIDOC-FGSKAQBVSA-M | |
| λ¹-rhodium(1+) bis(methanidylidyneoxidanium) (2Z)-4-oxopent-2-en-2-olate | |
| 258.04 | |
| Dicarbonylacetylacetonato rhodium(I) |
Safety and Handling
GHS H Statement
Flammable solid.
Toxic if swallowed.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
Risk of explosion if heated under confinement
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occ
GHS Signal Word: Danger
EINECSNumber : 238-947-9
RUO – Research Use Only