CAS RN 14874-82-9
CAS RN 14874-82-9
IUPAC Name:
λ¹-rhodium(1+) bis(methanidylidyneoxidanium) (2Z)-4-oxopent-2-en-2-olate
Synonyms:
λ¹-rhodium(1+) bis(carbon monoxide) (2Z)-4-oxopent-2-en-2-olate
Molecular Weight (g/mol):
258.03
Molecular Formula:
C7H7O4Rh
InChi Key:
BZCAWKOPWNIDOC-FGSKAQBVSA-M
SMILES:
[Rh+].[C-]#[O+].[C-]#[O+].C\C([O-])=C\C(C)=O
1
–
1
of
1
results
1
Dicarbonylacetylacetonato rhodium(I)
CAS: 14874-82-9 Molecular Formula: C7H7O4Rh Molecular Weight (g/mol): 258.03 MDL Number: MFCD00009884 InChI Key: BZCAWKOPWNIDOC-FGSKAQBVSA-M IUPAC Name: λ¹-rhodium(1+) bis(methanidylidyneoxidanium) (2Z)-4-oxopent-2-en-2-olate SMILES: [Rh+].[C-]#[O+].[C-]#[O+].C\C([O-])=C\C(C)=O