missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibutyltin Oxide 95.0+%, TCI America™
Supplier: TCI America D030525G
Specifications
| Dibutyltin Oxide | |
| White | |
| MFCD00001992 | |
| 3146 | |
| JGFBRKRYDCGYKD-UHFFFAOYSA-N | |
| dibutyl(oxo)tin | |
| 61221 | |
| ≥95.0% (W) |
| 818-08-6 | |
| C8H18OSn | |
| 25 g | |
| dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin | |
| CCCC[Sn](=O)CCCC | |
| 248.941 | |
| 248.94 | |
| Crystalline Powder |
Chemical Identifiers
| 818-08-6 | |
| 248.941 | |
| JGFBRKRYDCGYKD-UHFFFAOYSA-N | |
| 61221 | |
| CCCC[Sn](=O)CCCC |
| C8H18OSn | |
| MFCD00001992 | |
| dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin | |
| dibutyl(oxo)tin |
Safety and Handling
EINECSNumber : (2)-2031
RTECSNumber : WH7175000
TSCA : Yes