missing translation for 'onlineSavingsMsg'
Learn More

Dibutyltin Oxide 95.0+%, TCI America™

Catalog No. p-7126135
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D030525G 25 g
D0305100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D030525G Supplier TCI America Supplier No. D030525G
Only null left

Chemical Identifiers

CAS 818-08-6
Molecular Formula C8H18OSn
Molecular Weight (g/mol) 248.941
MDL Number MFCD00001992
InChI Key JGFBRKRYDCGYKD-UHFFFAOYSA-N
Synonym dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin
PubChem CID 61221
IUPAC Name dibutyl(oxo)tin
SMILES CCCC[Sn](=O)CCCC

Specifications

Color White
Quantity 25 g
UN Number 3146
Formula Weight 248.94
Percent Purity ≥95.0% (W)
Physical Form Crystalline Powder
Chemical Name or Material Dibutyltin Oxide
EINECSNumber (2)-2031
RTECSNumber WH7175000
TSCA Yes