missing translation for 'onlineSavingsMsg'
Learn More

Dibutyltin Dilaurate 95.0+%, TCI America™

Catalog No. p-7126134
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D030325G 25 g
D0303100G 100 g
D0303500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D030325G Supplier TCI America Supplier No. D030325G
Only null left

Chemical Identifiers

CAS 77-58-7
Molecular Formula C32H64O4Sn
MDL Number MFCD00008963

Specifications

Color Yellow
Physical Form Liquid
Quantity 25 g
UN Number 2788
Formula Weight 631.57
Percent Purity ≥95.0% (W)
Chemical Name or Material Dibutyltin Dilaurate
EINECSNumber (2)-2330
RTECSNumber WH7000000
TSCA Yes