accessibility menu, dialog, popup

UserName

CAS RN 77-58-7

IUPAC Name: dibutyltin dilaurate
Synonyms: dibutyltin dilaurate
Molecular Weight (g/mol): 633.59
Molecular Formula: C32H66O4Sn
InChi Key: XWVGXTYHPCKSLV-UHFFFAOYSA-N
SMILES: CCCC[SnH2]CCCC.CCCCCCCCCCCC(O)=O.CCCCCCCCCCCC(O)=O
Percent Purity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (6)
  • (1)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (6)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)