missing translation for 'onlineSavingsMsg'
Learn More

Dibutyltin Dichloride 97.0+%, TCI America™

Catalog No. D022325G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D022325G 25 g
D0223100G 100 g
D0223500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D022325G Supplier TCI America Supplier No. D022325G

Chemical Identifiers

CAS 683-18-1
Molecular Formula C8H18Cl2Sn
Molecular Weight (g/mol) 303.842
MDL Number MFCD00000518
InChI Key RJGHQTVXGKYATR-UHFFFAOYSA-L
Synonym dibutyltin dichloride, dibutyldichlorotin, di-n-butyltin dichloride, stannane, dibutyldichloro, dibutyltin chloride, dichlorodibutyltin, dibutyl dichloro stannane, dichlorodibutylstannane, dibutyltindichloride, dibutyl tin dichloride
PubChem CID 12688
IUPAC Name dibutyl(dichloro)stannane
SMILES CCCC[Sn](CCCC)(Cl)Cl

Specifications

Melting Point 42°C
Boiling Point 135°C
Quantity 25 g
UN Number 3146
Formula Weight 303.84
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dibutyltin Dichloride
EINECSNumber (2)-2331
RTECSNumber WH7100000
TSCA Yes