accessibility menu, dialog, popup

UserName

CAS RN 683-18-1

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 305.86
Molecular Formula: C8H20Cl2Sn
InChi Key: GSCBVGBIRKBMAC-UHFFFAOYSA-N
SMILES: Cl.Cl.CCCC[SnH2]CCCC
Boiling Point 
  • (3)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (7)
Percent Purity 
  • (3)
  • (3)
  • (2)
Physical Form 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (1)
  • (7)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)