missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibutyltin Diacetate 95.0+%, TCI America™
Supplier: TCI America D030225G
Specifications
| Dibutyltin Diacetate | |
| Yellow | |
| C12H26O4Sn | |
| 25 g | |
| LDZDQDDROJXUBO-UHFFFAOYSA-N | |
| 353.05 | |
| ≥95.0% (W) |
| 1067-33-0 | |
| 145°C | |
| MFCD00008697 | |
| 2922 | |
| CC(O)=O.CC(O)=O.CCCC[SnH2]CCCC | |
| 351.03 | |
| Liquid |
Chemical Identifiers
| 1067-33-0 | |
| 353.05 | |
| LDZDQDDROJXUBO-UHFFFAOYSA-N |
| C12H26O4Sn | |
| MFCD00008697 | |
| CC(O)=O.CC(O)=O.CCCC[SnH2]CCCC |
Safety and Handling
EINECSNumber : (2)-2330
RTECSNumber : WH6880000
TSCA : Yes