missing translation for 'onlineSavingsMsg'
Learn More

Dibutyldimethoxytin 95.0+%, TCI America™

Catalog No. D48365G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48365G 5 g
D483625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48365G Supplier TCI America Supplier No. D48365G

Chemical Identifiers

CAS 1067-55-6
Molecular Formula C10H24O2Sn
Molecular Weight (g/mol) 295.01
MDL Number MFCD00008345
InChI Key ZXDVQYBUEVYUCG-UHFFFAOYSA-N
PubChem CID 16683107
IUPAC Name dibutyl(dimethoxy)stannane
SMILES CCCC[Sn](CCCC)(OC)OC

Specifications

Color Yellow
Boiling Point 139°C
Quantity 5 g
UN Number 2788
Formula Weight 295.01
Percent Purity ≥95.0% (W)
Physical Form Liquid
Chemical Name or Material Dibutyldimethoxytin
EINECSNumber (2)-2318
TSCA Yes