accessibility menu, dialog, popup

UserName

H3COHSnOHCH3H3CCH3

CAS RN 1067-55-6

IUPAC Name: 1,1-dibutyl-O'1,O1-dimethylstannanediol
Synonyms: 1,1-dibutyl-O'1,O1-dimethylstannanediol
Molecular Weight (g/mol): 297.03
Molecular Formula: C10H26O2Sn
InChi Key: YZZBPBGRFCRIIZ-UHFFFAOYSA-N
SMILES: CO.CO.CCCC[SnH2]CCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results