missing translation for 'onlineSavingsMsg'
Learn More

Dibutylamine, 2 N in Toluene, Ricca Chemical

Catalog No. R25494004C
Change view
Click to view available options
Quantity:
4 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R25494004C 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R25494004C Supplier Ricca Chemical Company Supplier No. R25494004C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-92-2 , 108-88-3
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
InChI Key JQVDAXLFBXTEQA-UHFFFAOYSA-N
Synonym dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine
PubChem CID 8148
IUPAC Name N-butylbutan-1-amine
SMILES CCCCNCCCC

Specifications

CAS Min % 68.0, 30.08
CAS Max % 71.0, 31.3
Packaging Amber Glass Bottle
Solvent or Matrix Toluene
Quantity 4 L
Concentration or Composition Notes 2Normal
Chemical Name or Material Dibutylamine