missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Sulfoxide 96.0+%, TCI America™

Catalog No. D11735G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D11735G 5 g
D117325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D11735G Supplier TCI America Supplier No. D11735G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2168-93-6
Molecular Formula C8H18OS
Molecular Weight (g/mol) 162.291
MDL Number MFCD00002093
InChI Key LOWMYOWHQMKBTM-UHFFFAOYSA-N
Synonym dibutyl sulfoxide, butyl sulfoxide, n-butyl sulfoxide, dibutyl sulphoxide, di-n-butyl sulfoxide, butane, 1,1'-sulfinylbis, sulfoxide, dibutyl, di-n-butylsulfoxide, butane,1-butylsulfinyl, butylsulfinyl butane
PubChem CID 16568
IUPAC Name 1-butylsulfinylbutane
SMILES CCCCS(=O)CCCC

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 162.29
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Dibutyl Sulfoxide
RTECSNumber EK5700000
TSCA No