missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibutyl succinate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 453011000
| Quantity | 100mL |
|---|
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 141-03-7 | |
| 230.3 | |
| dibutyl succinate, tabutrex, succinic acid di-n-butyl ester, di-n-butyl succinate, tabatrex, dibutylsuccinate, butyl butanedioate, butanedioic acid, dibutyl ester, succinic acid dibutyl ester, dnbs | |
| CHEBI:82263 | |
| CCCCOC(=O)CCC(=O)OCCCC |
| C12H22O4 | |
| YUXIBTJKHLUKBD-UHFFFAOYSA-N | |
| 8830 | |
| dibutyl butanedioate |
Specifications
| Dibutyl succinate | |
| 0.9800g/mL | |
| 144°C | |
| C12H22O4 | |
| 100mL | |
| dibutyl succinate, tabutrex, succinic acid di-n-butyl ester, di-n-butyl succinate, tabatrex, dibutylsuccinate, butyl butanedioate, butanedioic acid, dibutyl ester, succinic acid dibutyl ester, dnbs | |
| YUXIBTJKHLUKBD-UHFFFAOYSA-N | |
| dibutyl butanedioate | |
| 8830 | |
| 230.3 |
| 141-03-7 | |
| 274.0°C | |
| 98.5% min. (GC) | |
| C4H9O2CH2CH2CO2C4H9 | |
| 0.98 | |
| Solubility in water: slightly. | |
| CCCCOC(=O)CCC(=O)OCCCC | |
| 230.3 | |
| CHEBI:82263 | |
| 99% |
Safety and Handling
EINECSNumber : 205-449-8
Promotions
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.