missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Phthalate, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111017871
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017871 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017871 Supplier Merck Emd Millipore Supplier No. PHR13661G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dibutyl Phthalate is a diester of phthalate, commonly synthesized by the condensation of the carboxy groups of phthalic acid with two molecules of 1-butanol. It is used as a plasticizer and film coating agent in solid pharmaceutical dosage forms.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 84-74-2
Vapor Pressure 1 mmHg (147°C)
Vapor Density 9.6 (vs. air)
Density 1.043 g/mL (at 25°C)
Linear Formula C6H4-1,2-[CO2(CH2)3 CH3]2
MDL Number MFCD00009441
Melting Point -35°C
Boiling Point 340°C
Synonym Dibutyl phthalate; n-Butyl phthalate; DBP; Phthalic acid dibutyl ester
Quantity 1 g
Formula Weight 278.34
Grade Certified Reference Material
Show More Show Less