missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Phosphite 95.0+%, TCI America™

Catalog No. d030025g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D030025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D030025G Supplier TCI America Supplier No. D030025G

Chemical Identifiers

CAS 1809-19-4
Molecular Formula C8H18O3P+
Molecular Weight (g/mol) 193.203
MDL Number MFCD00066633
InChI Key OSPSWZSRKYCQPF-UHFFFAOYSA-N
Synonym dibutyl phosphite, dibutyl phosphonate, phosphonic acid, dibutyl ester, di-n-butylphosphite, dibutoxyphosphine oxide, mobil dbhp, dibutyl hydrogen phosphonate, di-n-butyl hydrogen phosphite, dibutylfosfit, phosphorous acid, dibutyl ester
PubChem CID 6327349
IUPAC Name dibutoxy(oxo)phosphanium
SMILES CCCCO[P+](=O)OCCCC

Specifications

Color Colorless
Boiling Point 121°C
Quantity 25 g
Formula Weight 194.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dibutyl Phosphite
EINECSNumber (2)-2001
RTECSNumber HS6475000
TSCA Yes