missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Phosphate 97.0+%, TCI America™

Supplier:  TCI America P0260500G

 View more versions of this product

Catalog No. P0260500G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dibutyl Phosphate
-13°C
138°C
MFCD00015245
1718
JYFHYPJRHGVZDY-UHFFFAOYSA-N
dibutoxyphosphinic acid
7881
≥97.0% (T)
107-66-4
Yellow
C8H19O4P
500 g
dibutyl phosphate, di-n-butyl phosphate, phosphoric acid, dibutyl ester, dibutyl acid phosphate, di-n-butylphosphoric acid, dp 4 coupling agent, di-n-butyl hydrogen phosphate, unii-0072nn74tn, phosphoric acid di-n-butyl ester, phosphoric acid dibutyl ester
CCCCOP(O)(=O)OCCCC
210.21
210.21
Liquid

Chemical Identifiers

107-66-4
210.21
JYFHYPJRHGVZDY-UHFFFAOYSA-N
7881
CCCCOP(O)(=O)OCCCC
C8H19O4P
MFCD00015245
dibutyl phosphate, di-n-butyl phosphate, phosphoric acid, dibutyl ester, dibutyl acid phosphate, di-n-butylphosphoric acid, dp 4 coupling agent, di-n-butyl hydrogen phosphate, unii-0072nn74tn, phosphoric acid di-n-butyl ester, phosphoric acid dibutyl ester
dibutoxyphosphinic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1986

RTECSNumber : TB9605000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions