missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Phosphate 97.0+%, TCI America™

Catalog No. P026025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P026025G 25 g
P0260500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P026025G Supplier TCI America Supplier No. P026025G

Chemical Identifiers

CAS 107-66-4
Molecular Formula C8H19O4P
Molecular Weight (g/mol) 210.21
MDL Number MFCD00015245
InChI Key JYFHYPJRHGVZDY-UHFFFAOYSA-N
Synonym dibutyl phosphate, di-n-butyl phosphate, phosphoric acid, dibutyl ester, dibutyl acid phosphate, di-n-butylphosphoric acid, dp 4 coupling agent, di-n-butyl hydrogen phosphate, unii-0072nn74tn, phosphoric acid di-n-butyl ester, phosphoric acid dibutyl ester
PubChem CID 7881
IUPAC Name dibutoxyphosphinic acid
SMILES CCCCOP(O)(=O)OCCCC

Specifications

Melting Point -13°C
Color Yellow
Boiling Point 138°C
Quantity 25 g
UN Number 1718
Formula Weight 210.21
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Dibutyl Phosphate
EINECSNumber (2)-1986
RTECSNumber TB9605000
TSCA Yes