missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl phosphate, 97%

Catalog No. AC406645000
Change view
Click to view available options
Quantity:
250 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC406645000 500 g Glass bottle
AC406642500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC406645000 Supplier Thermo Scientific Chemicals Supplier No. 406645000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 107-66-4 | C8H19O4P | 210.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107-66-4
Molecular Formula C8H19O4P
Molecular Weight (g/mol) 210.21
MDL Number MFCD00015245
InChI Key JYFHYPJRHGVZDY-UHFFFAOYSA-N
Synonym dibutyl phosphate, di-n-butyl phosphate, phosphoric acid, dibutyl ester, dibutyl acid phosphate, di-n-butylphosphoric acid, dp 4 coupling agent, di-n-butyl hydrogen phosphate, unii-0072nn74tn, phosphoric acid di-n-butyl ester, phosphoric acid dibutyl ester
PubChem CID 7881
IUPAC Name dibutyl hydrogen phosphate
SMILES CCCCOP(O)(=O)OCCCC

Specifications

Acidity 259 to 267mg KOH/g
Melting Point -13.0°C
Density 1.0580g/mL
Boiling Point 250.0°C
Flash Point 180°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4270 to 1.4290
Linear Formula (CH3(CH2)3O)2P(O)OH
Quantity 500 g
Specific Gravity 1.058
Solubility Information Solubility in water: 17.2g/L
Viscosity 75 mPa.s (20°C)
Formula Weight 210.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dibutyl phosphate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water
EINECSNumber 203-509-8
RTECSNumber TB9605000
TSCA TSCA

RUO – Research Use Only