missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl N,N-Diethylcarbamoylmethylphosphonate [for Extraction of Lanthanides and Actinides], TCI America™

Catalog No. D15855ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15855ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D15855ML Supplier TCI America Supplier No. D15855ML

Chemical Identifiers

CAS 7439-68-1
Molecular Formula C14H30NO4P
Molecular Weight (g/mol) 307.37
MDL Number MFCD00015240
InChI Key SGGUHCSKZHSULP-UHFFFAOYSA-N
Synonym N,N-Diethylcarbamoylmethylphosphonic Acid Dibutyl Ester, DBDECMP
PubChem CID 2843612
IUPAC Name dibutyl [(diethylcarbamoyl)methyl]phosphonate
SMILES CCCCOP(=O)(CC(=O)N(CC)CC)OCCCC

Specifications

Color Yellow
Boiling Point 172°C
Quantity 5 mL
Formula Weight 307.37
Physical Form Liquid
Chemical Name or Material Dibutyl N,N-Diethylcarbamoylmethylphosphonate [for Extraction of Lanthanides and Actinides]