accessibility menu, dialog, popup

UserName

H3COPOONCH3CH3OCH3

CAS RN 7439-68-1

IUPAC Name: dibutyl [(diethylcarbamoyl)methyl]phosphonate
Synonyms: dibutyl (diethylcarbamoyl)methylphosphonate
Molecular Weight (g/mol): 307.37
Molecular Formula: C14H30NO4P
InChi Key: SGGUHCSKZHSULP-UHFFFAOYSA-N
SMILES: CCCCOP(=O)(CC(=O)N(CC)CC)OCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results