missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl L-(+)-Tartrate 98.0+%, TCI America™

Catalog No. p-7136024
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T000525G 25 g
T0005500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T000525G Supplier TCI America Supplier No. T000525G
Only null left

Chemical Identifiers

CAS 87-92-3
Molecular Formula C12H22O6
Molecular Weight (g/mol) 262.302
MDL Number MFCD00009443
InChI Key PCYQQSKDZQTOQG-NXEZZACHSA-N
Synonym 2r,3r-dibutyl 2,3-dihydroxysuccinate, dibutyl l-+-tartrate, dibutyl l-tartrate, unii-2d1v32if1e, dibutyl tartrate, +, l-+-tartaric acid di-n-butyl ester, dibutyl 2r,3r-2,3-dihydroxybutanedioate, butanedioic acid, 2,3-dihydroxy-2r,3r-, dibutyl ester, 1,4-dibutyl 2r,3r-2,3-dihydroxybutanedioate, dibutyll-+-tartrate
PubChem CID 6910
IUPAC Name dibutyl (2R,3R)-2,3-dihydroxybutanedioate
SMILES CCCCOC(=O)C(C(C(=O)OCCCC)O)O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 262.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dibutyl L-(+)-Tartrate
EINECSNumber (2)-1458
TSCA Yes