accessibility menu, dialog, popup

UserName

CAS RN 87-92-3

H3COO(R)OH(R)OHOOCH3

CAS RN 87-92-3

IUPAC Name: 1,4-dibutyl (2R,3R)-2,3-dihydroxybutanedioate
Synonyms: 1,4-dibutyl (2R,3R)-2,3-dihydroxybutanedioate
Molecular Weight (g/mol): 262.3
Molecular Formula: C12H22O6
InChi Key: PCYQQSKDZQTOQG-NXEZZACHSA-N
SMILES: CCCCOC(=O)[C@H](O)[C@@H](O)C(=O)OCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results