missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Fumarate 98.0+%, TCI America™

Catalog No. p-7129499
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F011625G 25 g
F0116500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F011625G Supplier TCI America Supplier No. F011625G
Only null left

Chemical Identifiers

CAS 105-75-9
Molecular Formula C12H20O4
Molecular Weight (g/mol) 228.288
MDL Number MFCD00065141
InChI Key JBSLOWBPDRZSMB-BQYQJAHWSA-N
Synonym dibutyl fumarate, butyl fumarate, staflex dbf, rc comonomer dbf, stafex dbf, fumaric acid, dibutyl ester, unii-cqm31z1nu9, dibutylmaleate, dibutyl e-but-2-enedioate, di-n-butyl fumarate
PubChem CID 5271570
IUPAC Name dibutyl (E)-but-2-enedioate
SMILES CCCCOC(=O)C=CC(=O)OCCCC

Specifications

Melting Point -18°C
Color Colorless
Boiling Point 285°C
Quantity 25 g
Formula Weight 228.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dibutyl Fumarate
EINECSNumber (2)-1099&(2)-1142
RTECSNumber LT1225000
TSCA Yes