accessibility menu, dialog, popup

UserName

CH3OOOOCH3(E)

CAS RN 105-75-9

IUPAC Name: 1,4-dibutyl (2E)-but-2-enedioate
Synonyms: dibutyl maleate
Molecular Weight (g/mol): 228.29
Molecular Formula: C12H20O4
InChi Key: JBSLOWBPDRZSMB-BQYQJAHWSA-N
SMILES: CCCCOC(=O)\C=C\C(=O)OCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results