missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibutyl Ether (stabilized with BHT) 99.0+%, TCI America™
$35.41 - $48.46
Chemical Identifiers
| CAS | 142-96-1 |
|---|---|
| Molecular Formula | C8H18O |
| Molecular Weight (g/mol) | 130.23 |
| MDL Number | MFCD00009461 |
| InChI Key | DURPTKYDGMDSBL-UHFFFAOYSA-N |
| Synonym | di-n-butyl ether, butyl ether, dibutyl ether, butane, 1,1'-oxybis, n-butyl ether, dibutyl oxide, butyl oxide, ether butylique, 1,1'-oxybis butane, di-n-butylether |
| PubChem CID | 8909 |
| IUPAC Name | 1-butoxybutane |
| SMILES | CCCCOCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B072125ML
|
TCI America
B072125ML |
25 mL |
Each for $35.41
|
|
|||||
|
B0721500ML
|
TCI America
B0721500ML |
500 mL |
Each for $48.46
|
|
|||||
Chemical Identifiers
| 142-96-1 | |
| 130.23 | |
| DURPTKYDGMDSBL-UHFFFAOYSA-N | |
| 8909 | |
| CCCCOCCCC |
| C8H18O | |
| MFCD00009461 | |
| di-n-butyl ether, butyl ether, dibutyl ether, butane, 1,1'-oxybis, n-butyl ether, dibutyl oxide, butyl oxide, ether butylique, 1,1'-oxybis butane, di-n-butylether | |
| 1-butoxybutane |
Specifications
| 142-96-1 | |
| Colorless | |
| C8H18O | |
| 25 mL | |
| di-n-butyl ether, butyl ether, dibutyl ether, butane, 1,1'-oxybis, n-butyl ether, dibutyl oxide, butyl oxide, ether butylique, 1,1'-oxybis butane, di-n-butylether | |
| CCCCOCCCC | |
| 130.23 | |
| 130.23 | |
| Liquid |
| -98°C | |
| 141°C | |
| MFCD00009461 | |
| 1149 | |
| DURPTKYDGMDSBL-UHFFFAOYSA-N | |
| 1-butoxybutane | |
| 8909 | |
| ≥99.0% (GC) | |
| Dibutyl Ether (stabilized with BHT) |
Safety and Handling
EINECSNumber : (2)-0363
RTECSNumber : EK5425000
TSCA : Yes