missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Ether (stabilized with BHT) 99.0+%, TCI America™

Supplier:  TCI America B0721500ML

 View more versions of this product

Catalog No. B0721500ML

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dibutyl Ether (stabilized with BHT)
-98°C
141°C
MFCD00009461
1149
DURPTKYDGMDSBL-UHFFFAOYSA-N
1-butoxybutane
8909
≥99.0% (GC)
142-96-1
Colorless
C8H18O
500 mL
di-n-butyl ether, butyl ether, dibutyl ether, butane, 1,1'-oxybis, n-butyl ether, dibutyl oxide, butyl oxide, ether butylique, 1,1'-oxybis butane, di-n-butylether
CCCCOCCCC
130.23
130.23
Liquid

Chemical Identifiers

142-96-1
130.23
DURPTKYDGMDSBL-UHFFFAOYSA-N
8909
CCCCOCCCC
C8H18O
MFCD00009461
di-n-butyl ether, butyl ether, dibutyl ether, butane, 1,1'-oxybis, n-butyl ether, dibutyl oxide, butyl oxide, ether butylique, 1,1'-oxybis butane, di-n-butylether
1-butoxybutane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0363

RTECSNumber : EK5425000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions