missing translation for 'onlineSavingsMsg'
Learn More

Dibromopyromellitic Dianhydride 98.0+%, TCI America™

$557.35 - $1660.87

Chemical Identifiers

CAS 24848-78-0
Molecular Formula C10Br2O6
Molecular Weight (g/mol) 375.91
MDL Number MFCD01100205
InChI Key OYIHYCLAXDDYFC-UHFFFAOYSA-N
Synonym 3,6-Dibromobenzene-1,2,4,5-tetracarboxylic 1,2:4,5-Dianhydride
PubChem CID 630455
IUPAC Name dibromo-5,11-dioxatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),8-triene-4,6,10,12-tetrone
SMILES BrC1=C2C(=O)OC(=O)C2=C(Br)C2=C1C(=O)OC2=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D49471G
View Documents
TCI America
D49471G
1 g
Each for $557.35
Only null left
 
D49475G
View Documents
TCI America
D49475G
5 g
Each for $1,660.87
Only null left
 
Description

Description

Specifications

Chemical Identifiers

24848-78-0
375.91
OYIHYCLAXDDYFC-UHFFFAOYSA-N
630455
BrC1=C2C(=O)OC(=O)C2=C(Br)C2=C1C(=O)OC2=O
C10Br2O6
MFCD01100205
3,6-Dibromobenzene-1,2,4,5-tetracarboxylic 1,2:4,5-Dianhydride
dibromo-5,11-dioxatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),8-triene-4,6,10,12-tetrone

Specifications

24848-78-0
C10Br2O6
1 g
OYIHYCLAXDDYFC-UHFFFAOYSA-N
dibromo-5,11-dioxatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),8-triene-4,6,10,12-tetrone
630455
≥98.0% (HPLC)
Dibromopyromellitic Dianhydride
275°C
MFCD01100205
3,6-Dibromobenzene-1,2,4,5-tetracarboxylic 1,2:4,5-Dianhydride
BrC1=C2C(=O)OC(=O)C2=C(Br)C2=C1C(=O)OC2=O
375.91
375.91
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'