missing translation for 'onlineSavingsMsg'
Learn More

Dibromomethane, MilliporeSigma™ Supelco™

Catalog No. 111020487
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020487 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020487 Supplier Merck Emd Millipore Supplier No. 47799

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Environmental

Specifications

Packaging Ampule
CAS 74-95-3
Vapor Pressure 34.9 mmHg (20°C)
Vapor Density 6.05 (vs. air)
Density 2.477 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH2 Br2
MDL Number MFCD00000168
Melting Point -52°C
Boiling Point 96°C to 98°C
Synonym Methylene bromide
Type Organic Pollutant Standard
Quantity 1000 mg
Formula Weight 173.83
Grade Analytical Standard
Show More Show Less