missing translation for 'onlineSavingsMsg'
Learn More

Dibromomethane, 99%

Catalog No. AC112831000
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC112831000 100 mL Glass bottle
AC112835000 500 mL Glass bottle
AC112830010 1 L Glass bottle
AC112830025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC112831000 Supplier Thermo Scientific Chemicals Supplier No. 112831000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 74-95-3 | CH2Br2 | 173.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 74-95-3
Molecular Formula CH2Br2
Molecular Weight (g/mol) 173.83
MDL Number MFCD00000168
InChI Key FJBFPHVGVWTDIP-UHFFFAOYSA-N
Synonym methylene bromide, methane, dibromo, methylene dibromide, dibromomethylene, rcra waste number u068, dibrommethan, methylenbromid, dibromo-methane, ccris 939, rcra waste no. u068
PubChem CID 3024
ChEBI CHEBI:47077
IUPAC Name dibromomethane
SMILES C(Br)Br

Specifications

Melting Point -52.0°C
Color Colorless to Brown
Density 2.4700g/mL
Boiling Point 96.0°C to 98.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5400 to 1.5420
Linear Formula CH2Br2
Quantity 100 mL
Beilstein 01,67
Fieser 11,337; 12,322; 16,393
Merck Index 15,6133
Specific Gravity 2.47
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: miscible with alcohol, ether, acetone and, chloroform
Formula Weight 173.83
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Dibromomethane, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful to aquatic life with long lasting effects.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 200-824-2
RTECSNumber PA7350000
TSCA TSCA

RUO – Research Use Only