Learn More
Dibromomalonamide, 98+%
CAS: 73003-80-2 | C3H4Br2N2O2 | 259.89 g/mol
$110.33 - $661.99
Chemical Identifiers
| CAS | 73003-80-2 |
|---|---|
| Molecular Formula | C3H4Br2N2O2 |
| Molecular Weight (g/mol) | 259.89 |
| MDL Number | MFCD00014803 |
| InChI Key | SWHQVMGRXIYDSF-UHFFFAOYSA-N |
| PubChem CID | 175375 |
| SMILES | NC(=O)C(Br)(Br)C(N)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2231509
|
Thermo Scientific Chemicals
B2231509 |
10 g |
Each for $110.33
|
|
|||||
|
AAB2231514
|
Thermo Scientific Chemicals
B2231514 |
25 g |
Each for $215.37
|
|
|||||
|
AAB2231522
|
Thermo Scientific Chemicals
B2231522 |
100 g |
Each for $661.99
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
ApplicationsDibromomalonamide is an antimicrobial agent used in sugar mills, a compound for lipophilic fluid systems for fabric care. It is also used to produce 5-thioxo-[1,3]dithiolo[4,5-d][1,3]dithiole-2,2-dicarboxylic acid diamide.
Solubility
Soluble in water.
Notes
Moisture & light sensitive. Store away from oxidizing agent.
Chemical Identifiers
| 73003-80-2 | |
| 259.89 | |
| SWHQVMGRXIYDSF-UHFFFAOYSA-N | |
| NC(=O)C(Br)(Br)C(N)=O |
| C3H4Br2N2O2 | |
| MFCD00014803 | |
| 175375 |
Specifications
| 73003-80-2 | |
| C3H4Br2N2O2 | |
| 10 g | |
| Light and moisture sensitive | |
| Soluble in water. | |
| NC(=O)C(Br)(Br)C(N)=O | |
| 259.89 | |
| 259.89 | |
| Dibromomalonamide |
| 202°C to 207°C | |
| MFCD00014803 | |
| 1770873 | |
| dibromomalonamide, 2,2-dibromomalonamide, dibromopropanediamide, propanediamide, 2,2-dibromo, dibromo malonamide-13c3, dibromo malonamide, dibromomalonic diamide, dibromomalonic acid diamide, propanediamide,2,2-dibromo, 2,2-bis bromanyl propanediamide | |
| SWHQVMGRXIYDSF-UHFFFAOYSA-N | |
| dibromopropanediamide | |
| 175375 | |
| ≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 277-205-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only